NSI Lab Solutions MIC-002 Quantitative Standard Plate Count Drinking Water Proficiency Test

Representative image only.

NSI Lab Solutions MIC-002 Quantitative Standard Plate Count Drinking Water Proficiency Test

NSI Lab Solutions | Item: 0500459

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Includes

Sterile hydration fluid.

Key Features

  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • Supplied in duplicate for convenience and value
  • Report quarterly quantitative results
  • Pellet contains heterotrophic bacteria at 5 to 500 MPN/CFU/mL

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental microbiology drinking water proficiency tests are designed for quantitative and/or qualitative identification of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismEnterococci
  • Organism typeBacteria
  • Packaging2/PK
  • Product TypeDrinking Water Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Standard Plate Count Drinking Water Proficiency Test
Reviews
Q&A