NSI Lab Solutions FMPT-025DB Qualitative E. coli O157:H7 with Dairy Matrix PT Express™ Set; 3/PK

Representative image only.

NSI Lab Solutions FMPT-025DB Qualitative E. coli O157:H7 with Dairy Matrix PT Express™ Set; 3/PK

NSI Lab Solutions | Item: 0500364

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

Three jars of sterile skim milk powder matrix and three individually packaged lyophilized microorganism pellets.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For qualitative identification of E. coli O157:H7
  • Designed to mimic typical FDA BAM and USDA MLG sample handling procedures
  • Set of three

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results. Your PT report is available within 48 hours once you submit your results online.

Food Microbiology PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Each three-sample set contains 3 x 25 g of sterile skim milk powder matrix and three individually packaged lyophilized microorganism pellets. At least one of three of the sample pellets will be positive for E. coli O157:H7. To use, transfer 25 g of sterile matrix to stomacher bag, add 225 mL of enrichment broth (not included), then add a single sample pellet to the stomacher bag. Process, analyze, then report present or absent for each sample in the set. Three different series available per study.

Specifications
  • OrganismE. coli
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CMatrix: store at room temperature
  • StrainO157:H7
  • DescriptionQualitative E. coli O157:H7 with Dairy Matrix PT Express™ Set; 3/PK
Reviews
Q&A