Five Snap-Stick™ inoculating swabs and five 4" x 4" sterile square petri dishes.
Key Features
Fast turnaround—PT reports available within 48 hours!
Available year-round
Convenient online reporting with PT Datalink
Submit PT report to accreditation agencies at no additional charge
Hydration fluid dissolves culture pellet in less than a minute
Glass-free culture swab device ensures user safety
ISO 17043 accredited
A five-sample proficiency testing panel
For qualitative identification of E. coli O157:H7
More About this Item
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
Each qualitative food microbiology proficiency test comes with five Snap-Stick™ inoculated swabs and five 4” x 4” sterile square petri dishes. To activate the microorganism, simply apply pressure to the plastic stem valve and release the hydration fluid to dissolve the microorganism pellet. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Once dissolved, remove the swab from the tube and swipe across the square petri dish base. At least 2 of 5 samples swabs will test positive for the target microorganism; acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives.
Food Microbiology PT Express Swabs may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Specifications
OrganismE. coli
Organism typeBacteria
Packaging5/PK
Product TypeFood Microbiology PT Express™
Shelf Life Months (months)24
Shipping requirementFrozen ice packs
Storage requirementRefrigerate between 2 and 8°C
StrainO157:H7
DescriptionQualitative E. coli O157:H7 Environmental Swab PT Express™; 5/PK