NSI Lab Solutions FMPT-013B Qualitative Campylobacter with Water Matrix PT Express™ Set; 3/PK

Representative image only.

NSI Lab Solutions FMPT-013B Qualitative Campylobacter with Water Matrix PT Express™ Set; 3/PK

NSI Lab Solutions | Item: 0500359

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

Hydration fluid.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For qualitative identification of Campylobacter spp.
  • Water matrix
  • Set of three

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results. Your PT report is available within 48 hours once you submit your results online.

Food Microbiology PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

This three-sample set contains C. jejuni or C. coli plus nonpathogenic/non-campylobacter facultative anaerobes. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. Report present or absent for Campylobacter spp. for each sample in the set.

Specifications
  • Organism C. jejuni or C. coli
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQualitative Campylobacter in Water Matrix PT Express™ Set; 3/PK
Reviews
Q&A