NSI Lab Solutions FMPT-009M Qualitative STEC with Meat Matrix Proficiency Test; 2/PK

Representative image only.

NSI Lab Solutions FMPT-009M Qualitative STEC with Meat Matrix Proficiency Test; 2/PK

NSI Lab Solutions | Item: 6120375

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Includes

Hydration fluid and meat matrix.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • For qualitative identification of Shiga toxin-producing E. coli (STEC)
  • Meat matrix
  • Supplied in duplicate for convenience and value

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. Receive your final reports within 21 days of the study close date.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

A single sample conveniently supplied in duplicate containing at least one of six STECs for identification. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismShiga toxin-producing E. coli (STEC)
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid and matrix: store at room temperature
  • StrainO26, O45, O103, O111, O121, or O145
  • DescriptionQualitative STEC with Meat Matrix Proficiency Test; 2/PK
Reviews
Q&A