NSI Lab Solutions FC-888 Total Milk Allergen Reference Standard, Blank, 50 grams

Representative image only.

NSI Lab Solutions FC-888 Total Milk Allergen Reference Standard, Blank, 50 grams

NSI Lab Solutions | Item: 0500483

Validate and evaluate samples for common food allergens with quality reference standards

Call For Price

Call For Price
Product Description

Key Features

  • Improve safety with blank, high, and low reference materials
  • Validate the accuracy of your allergen testing methods
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 and ISO 9001 requirements
  • Inert Blank Matrix certified <1 ppm Total Milk by both Neogen Veratox Milk and R-Biopharm R4652

More About this Item

Food allergen reference material standards are used primarily for quality control and validation purposes in food testing applications including calibration, validation, and quality control testing. These reference materials are crucial in demonstrating testing procedures are sensitive and specific enough to detect allergens at levels relevant to consumer safety. By testing reference materials with known allergen concentrations, labs can verify the accuracy, precision, and reliability of their methods. Allergens are available in blank, high, and low concentrations.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

Specifications
  • Concentration <1 ppm total milk
  • OrganismTotal Milk
  • Organism typeNeogen Veratox Milk and R-Biopharm R4652
  • PackagingAmber glass jar
  • Product TypeBlank
  • Shelf Life Months (months)24
  • Storage requirementRoom temperature
  • Weight g (g)50
  • DescriptionTotal Milk Allergen Reference Standard, Blank, 50 grams
Documents & Certificates
Reviews
Q&A