NSI Lab Solutions CMPT-040 Quantitative Yeast/Mold in Hemp Proficiency Test

Representative image only.

NSI Lab Solutions CMPT-040 Quantitative Yeast/Mold in Hemp Proficiency Test

NSI Lab Solutions | Item: 0500144

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Includes

Two bottles of hydration fluid.

Key Features

  • Designed for quantitative determination
  • Two studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Applicable to cultural techniques
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • For quantitative determination of yeast/mold in the range of 500 to 50,000 CFU/pellet
  • Supplied as hemp inoculated with yeast and/or mold

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismYeast, mold
  • Organism typeFungi
  • Product TypeHemp Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Yeast/Mold in Hemp Proficiency Test; 2/PK
Reviews
Q&A