NSI Lab Solutions CMPT-037B Quantitative Coliforms/E. coli in Hemp PT Express™

Representative image only.

NSI Lab Solutions CMPT-037B Quantitative Coliforms/E. coli in Hemp PT Express™

NSI Lab Solutions | Item: 0500107

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

Two bottles of hydration fluid.

Key Features

  • Designed for quantitative determination
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • For quantitative determination of total coliforms and E. coli in the range of 500 to 500,000 CFU/pellet
  • Supplied as hemp inoculated with several BSL-2 aerobic organisms

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results. Your PT report is available within 48 hours once you submit your results online.

These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.

Hemp PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • Biosafety requirementBSL-2
  • OrganismTotal Coliforms, E. coli
  • Organism typeBacteria
  • Product TypeHemp PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Coliforms/E. coli in Hemp PT Express™
Reviews
Q&A