NSI Lab Solutions CMPT-035B Qualitative E. coli O157:H7 in Hemp PT Express™; 5/PK

Representative image only.

NSI Lab Solutions CMPT-035B Qualitative E. coli O157:H7 in Hemp PT Express™; 5/PK

NSI Lab Solutions | Item: 0500065

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • A five-sample set inoculated in hemp
  • Qualitative test for E. coli O157:H7

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

For the qualitative hemp proficiency tests, at least 2 of 5 samples will test positive for the target microorganism. Acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives. Your PT report is available within 48 hours once you submit your results online. These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.

Hemp PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • OrganismE. coli
  • Organism typeBacteria
  • Packaging5/PK
  • Product TypeHemp PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainO157:H7
  • DescriptionQualitative E. coli O157:H7 in Hemp PT Express; 5/PK
Reviews
Q&A