NSI Lab Solutions CMPT-031B Qualitative Aspergillus Molds in Hemp PT Express™; 5/PK

Representative image only.

NSI Lab Solutions CMPT-031B Qualitative Aspergillus Molds in Hemp PT Express™; 5/PK

NSI Lab Solutions | Item: 0500053

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • A five-sample set inoculated in hemp
  • Qualitative test for Aspergillus molds (A. flavus, A. fumigatus, A. niger, A. terreus)

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

For the qualitative hemp proficiency tests, at least 2 of 5 samples will test positive for the target microorganism. Acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives. Your PT report is available within 48 hours once you submit your results online. These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.

Hemp PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • OrganismA. flavus, A. fumigatus, A. niger, A. terreus
  • Organism typeFungi
  • Packaging5/PK
  • Product TypeHemp PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • DescriptionQualitative Aspergillus Molds in Hemp PT Express™; 5/PK
Reviews
Q&A