NSI Lab Solutions CMPT-029 Qualitative STEC in Edible Proficiency Test; 5/PK

Representative image only.

NSI Lab Solutions CMPT-029 Qualitative STEC in Edible Proficiency Test; 5/PK

NSI Lab Solutions | Item: 0500087

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Key Features

  • Two studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • A five-sample set inoculated in edible
  • Qualitative test for Shiga toxin-producing E. coli, STEC (O26, O45, O103, O111, O121, or O14)

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

For the qualitative hemp proficiency tests, at least 2 of 5 samples will test positive for the target microorganism. Acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives. These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismShiga toxin-producing E. coli (STEC)
  • Organism typeBacteria
  • Packaging5/PK
  • Product TypeHemp Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainO26, O45, O103, O111, O121, or O14
  • DescriptionQualitative STEC in Edible Proficiency Test; 5/PK
Reviews
Q&A