NSI Lab Solutions 51-00053 A. brasiliensis NCPF 2275 Pharmaceutical Culture, USP <51>; 10/PK

Representative image only.

NSI Lab Solutions 51-00053 A. brasiliensis NCPF 2275 Pharmaceutical Culture, USP <51>; 10/PK

NSI Lab Solutions | Item: 0500171

Comply with guidelines set by the USP for antimicrobial effectiveness testing

Call For Price

Call For Price
Product Description

Includes

Ten 1.5 mL vials of peptone buffer.

Key Features

  • When hydrated, activity will yield at least 1 x 108 CFU/mL
  • Stable for up to 8 hours after hydration
  • Includes sterile peptone buffer for convenience
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Equivalent to WDCM 00053 and ATCC 16404

More About this Item

Designed to conform with USP <51> specifications, culture pellet is manufactured from USP specified pure organism strain. For your convenience, each pack includes ten culture pellets individually vacuum sealed in vials and ten 1.5 mL vials of sterile peptone buffer. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Once hydrated in the supplied peptone buffer, these samples are stable for up to 8 hours. Extra activity listed on Certificate of Analysis (CoA). Store in the freezer.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

ATCC is a registered trademark of American Type Culture Collection.
NCTC and NCPF reference cultures obtained from the National Collection of Type Cultures and National Collection of Pathogenic Fungi, two of the culture collections of the UK Health Security Agency.
WDCM reference cultures obtained from the WFCC-MIRCEN World Data Centre for Organisms.

Specifications
  • OrganismA. brasiliensis
  • Packaging10/PK
  • Product TypeUSP <51> Pharmaceutical Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCPF 2275, WDCM 00053, ATCC 16404
  • DescriptionA. brasiliensis NCPF 2275 Pharmaceutical Culture, USP <51>; 10/PK
Reviews
Q&A