NSI Lab Solutions 10662-10 P. aeruginosa High Range NCTC 10062 Environmental Culture; 10/PK
Representative image only.
NSI Lab Solutions 10662-10 P. aeruginosa High Range NCTC 10062 Environmental Culture; 10/PK
NSI Lab Solutions | Item: 0500291
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
Call For Price
Call For Price
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 10
More About this Item
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
Use P. aeruginosa NCTC 12951 culture as a negative control for water coliform analysis or as a positive quantitative control for water pseudomonas analysis. The Certificate of Analysis (CoA) lists value assignments for MPN methods and plate count methods. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
NCTC reference cultures obtained from the National Collection of Type Cultures.
Specifications
  • OrganismP. aeruginosa
  • Packaging10/PK
  • Product TypeEnvironmental Single Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 12951
  • DescriptionP. aeruginosa High Range NCTC 10062 Environmental Culture; 10/Pk