Mettler Toledo Micro pH Electrode; S7
Representative image only.
Mettler Toledo Micro pH Electrode; S7
Mettler Toledo | Mfr: 51343160| Item: 3590141
Expertly-engineered probes to cover a wide range of applications
Call For Price
Call For Price
Buy It Together
Current Item
Mettler Toledo Micro pH Electrode; S7
Mettler Toledo Micro pH Electrode; S7
3590141
Call For Price
+
Environmental Express T80WL Portable White Light Turbidity Meter Kit, EPA Compliant
3563521
£997.55GBP / Each
£1,197.06GBP (incl VAT)
+ + +
Cole-Parmer Adjustable-Volume Pipettor, 2000 to 10000 µL
2450110
£155.15GBP / Each
£186.18GBP (incl VAT)
+ =
Total Price 1614.81 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cable not included⌡ARGENTHAL™ reference system
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.
Specifications
  • ApplicationsSmall volumes and containers
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ATC Element
  • ConnectorS7
  • Diameter (mm)3
  • Length (mm)60
  • DescriptionpH Electrode; S7