Meiji Techno Microscope Stage Micrometer, 0.04"/0.001" Scale
Representative image only.
Meiji Techno Microscope Stage Micrometer, 0.04"/0.001" Scale
Meiji Techno | Mfr: MA286| Item: 4840481
Make quick and accurate measurements of your specimen
£107.86GBP / Each
£129.43GBP (incl VAT)
£107.86GBP / Each
£129.43GBP (incl VAT)
Buy It Together
Current Item
Meiji Techno Microscope Stage Micrometer, 0.04"/0.001" Scale
Meiji Techno Microscope Stage Micrometer, 0.04"/0.001" Scale
4840481
£107.86GBP / Each
£129.43GBP (incl VAT)
+
Meiji Techno Microscope Eyepieces, Super Widefield, 10X, 23 mm Field of View; Pair
4840258
£174.23GBP / Pair of 1
£209.08GBP (incl VAT)
=
Total Price 282.09 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily count or measure sample when observing samples under a microscope
  • Limited Lifetime Warranty
More About this Item
Microscope micrometers, also referred to as reticles, are commonly used for measuring or counting specimens. Eyepiece micrometers are small glass discs with markings on them. The micrometer is mounted in one of the two eyepieces and superimposes an image of the markings over the image of the specimen. Stage micrometers are slides with markings of a known dimension that, when viewed under the microscope, can be used to calibrate eyepiece micrometers.
Limited Lifetime Warranty
Specifications
  • Width (InchShort)0.04
  • Slide TypeStage Micrometer
  • Size0.04"/0.001" scale
  • DescriptionMicroscope Stage Micrometer, 0.04"/0.001" Scale
  • WarrantyLTD Life Time