Labconco Xpert 3961620 6-ft Bulk Powder Filtered System, Left Side Access; 230 VAC

Representative image only.

Labconco Xpert 3961620 6-ft Bulk Powder Filtered System, Left Side Access; 230 VAC

Labconco Xpert | Mfr: 3961620| Item: 3350915

Protect yourself during powder transfer operations

Call For Price

Call For Price

Buy It Together

Current Item

Labconco Xpert 3961620 6-ft Bulk Powder Filtered System, Left Side Access; 230 VAC
Labconco Xpert 3961620 6-ft Bulk Powder Filtered System, Left Side Access; 230 VAC
3350915

Call For Price
+ =
Total Price 592.20 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Stainless steel work surface with front spill trough to catch spills
  • Push-button hydraulic lift for source container
  • 1 year warranty

More About this Item

XPert® bulk powder filter systems protect you from hazardous airborne particulates and fumes generated during powder transfer operations. Systems feature built-in blowers with dynamically balanced, vibration-isolated impellers with speed control, pre-wired fluorescent lamp with switch, and exhaust HEPA filter efficient on particled 0.3 microns. Models also available with center and right-side source container access openings. Call our Application Specialists for information regarding enclosures and filtered stations.
What’s included: transfer caddy with 3"-dia casters, 2"-square tubular steel base with leveling feet, exhaust HEPA filter, built-in blower with speed control, pre-wired fluorescent lamp and switches for blower and light, digital airflow

1 Year WarrantyETL ListedISO9001:2000 Certified Supplier
Specifications
  • Exterior Width (in)72
  • Exterior Height (in)37
  • Exterior Depth (in)83.3
  • Power (VAC)230
  • Power (Hz)50
  • Description6' Bulk Powder Filtered System 230V Left side access
  • Warranty1 Year
Reviews
Q&A