Labconco RXPert Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V

Representative image only.

Labconco RXPert Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V

Labconco | Mfr: 3971486| Item: 3252151

Keep hazardous powders and particulates contained

Call For Price

Call For Price

Buy It Together

Current Item

Labconco RXPert  Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V
Labconco RXPert Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V
3252151

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Includes

8-ft (2.4-m) power cord.

Key Features

  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • Outside ducting optional
  • 1 year warranty

More About this Item

The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.


All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.


Double-Filtered Enclosures feature redundant filters in series, so do not require exhausting to the outside while maintaining USP <800> compliance. All models require a supporting base (order separately).


Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.

1 Year Warranty
Specifications
  • Hood Width (in)48
  • Exterior Width (in)48
  • Exterior Height (in)58.8125
  • Exterior Depth (in)29
  • Interior Width (in)45.625
  • Interior Height (in)32
  • Interior Depth (in)23.4375
  • Power (VAC)115
  • Power (Hz)60
  • Sash opening (in)12
  • Exhaust Volume (CFM)380
  • Shipping Weight (lbs)520
  • Shipping Weight (kg)236
  • DescriptionTall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V
  • Warranty1 Year
Reviews
Q&A