Labconco FreeZone Freeze Drying Manifold, 14 Ports; Console FD
Representative image only.
Labconco FreeZone Freeze Drying Manifold, 14 Ports; Console FD
Labconco | Mfr: 7201900| Item: 0335310
Manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks
Call For Price
Call For Price
Buy It Together
Current Item
Labconco FreeZone Freeze Drying Manifold, 14 Ports; Console FD
Labconco FreeZone Freeze Drying Manifold, 14 Ports; Console FD
0335310
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
one latching pin.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” adapters for quick connection of flasks
  • Compatible with 6-, 12-, and 18-L FreeZone Freeze Dry Consoles
  • 1 year warranty
More About this Item
Stainless steel manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks.
1 Year Warranty
Specifications
  • DescriptionFreeze Drying Manifold, 14 Ports; Console FD