Labconco FreeZone Fast-Freeze Flask, Amber; 600 mL
Representative image only.
Labconco FreeZone Fast-Freeze Flask, Amber; 600 mL
Labconco | Mfr: 7542801| Item: 0335329
£239.61GBP / Each
£287.53GBP (incl VAT)
£239.61GBP / Each
£287.53GBP (incl VAT)
Buy It Together
Current Item
Labconco FreeZone Fast-Freeze Flask, Amber; 600 mL
Labconco FreeZone Fast-Freeze Flask, Amber; 600 mL
0335329
£239.61GBP / Each
£287.53GBP (incl VAT)
+
Labconco Fast-Freeze™ 7540800 Clear Glass Drying Flask; 600 mL
0335305
£402.78GBP / Each
£483.34GBP (incl VAT)
+
Labconco FreeZone Standard Freeze Drying Chamber, 12 Ports, SS
0334601
£2,660.39GBP / Each
£3,192.47GBP (incl VAT)
=
Total Price 3302.78 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Withstands extreme temperatures and high vacuum
  • Amber-colored glass protects light-sensitive samples
  • Flat bottom and wide mouth design provides easy loading and cleaning
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
Specifications
  • Capacity (Liters)0.6
  • Height (InchShort)5 21/64
  • DescriptionFast-Freeze Flask, Amber, Flat Bottom, 600 mL