Jenco Ph/Orp Controller Meter Only
Representative image only.
Jenco Ph/Orp Controller Meter Only
Jenco | Mfr: 3671| Item: 0565600
Features high and low overrange lights for quick indication of out of parameter readings
Call For Price
Call For Price
Buy It Together
Current Item
Jenco Ph/Orp Controller Meter Only
Jenco Ph/Orp Controller Meter Only
0565600
Call For Price
+ =
Total Price 298.59 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 6 month warranty
More About this Item
Panel-mount controller includes many features usually found on higher priced models. Unit features recorder output, automatic temperature compensation, and high/low overrange lights.
Adjust set points, slope, and calibration via recessed front-panel potentiometers. Push buttons let you display current process value, high or low set point, or temperature. Select pH or ORP mode via an internal switch.
Two SPDT relays control external devices. Relays are protected by a RC suppression circuit. Order disposable electrodes below.
6 Month Warranty
Specifications
  • Control TypeOn/Off
  • Display TypeLED
  • DescriptionPh/Orp Controller Meter Only