Idex VHP Union Body, ZDV, Stainless Steel, 0.020" ID, 1/16" Tubing, 10-32 Coned
Representative image only.
Idex VHP Union Body, ZDV, Stainless Steel, 0.020" ID, 1/16" Tubing, 10-32 Coned
Idex | Mfr: U-402-01| Item: 0202482
Zero dead volume unions ensure accurate analysis
£25.25GBP / Each
£30.30GBP (incl VAT)
£25.25GBP / Each
£30.30GBP (incl VAT)
Product Description
Key Features
  • Precision machined from 316 stainless steel to withstand high pressures
  • Securely join fluid pathway components
  • Holds tight connection even through vibration
  • VHP ZDV union body
  • 10-32 coned configuration
  • 0.020" thru-hole diameter
  • Swept volume of 0.13 µL
  • 1 year warranty
More About this Item
These Idex® VHP stainless steel ZDV unions provide a tight, secure connection in your chromatography fluid path.
1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • DescriptionVHP Union Body, ZDV, SS, 0.020" ID, 1/16" Tubing, 10-32 Coned; 1/EA