Idex VacuTight™ Ferrule, Red ETFE, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
Representative image only.
Idex VacuTight™ Ferrule, Red ETFE, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA
Idex | Mfr: P-940| Item: 0201763
Make dependable connections under vacuum and low pressure conditions
£1.45GBP / Each
£1.74GBP (incl VAT)
£1.45GBP / Each
£1.74GBP (incl VAT)
Product Description
Key Features
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
  • For 1/8" OD tubing
  • Works with M6 or 1/4-28 flat-bottom ports
  • 1/EA
  • 1 year warranty
More About this Item
VacuTight fittings are ideal for high throughput screening and other automated liquid-handling applications due to their small size and dependable, airtight connections in low pressure and vacuum environments. The flat-bottom ferrule configuration impedes tubing inner diameter reduction and overcompression that can be a problem with many coned fittings.
All VacuTight nuts must be used only with VacuTight ferrules.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • ColorRed
  • DescriptionFerrule, Red ETFE, 1/8" OD Tubing, 1/4-28 or M6 Flat Bottom; 1/EA