Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
Representative image only.
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
Idex | Mfr: XP-335| Item: 0202099
Short head is ideal for tight-speace applications
£8.96GBP / Each
£10.75GBP (incl VAT)
£8.96GBP / Each
£10.75GBP (incl VAT)
Buy It Together
Current Item
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
0202099
£8.96GBP / Each
£10.75GBP (incl VAT)
+ +
Cole-Parmer VapLock™ Solvent Delivery Cap, two 1/4"-28 ports, GL45; 1/ea
1201803
£44.91GBP / Each
£53.89GBP (incl VAT)
=
Total Price 57.33 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Yellow ETFE ferrule 02019-82.
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural PEEK
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8