HTL 7905 Labmate Pro Starter Kit with LMP10, LMP20, LMP200 & LMP1000 Pipettors and Accessories
Representative image only.
HTL 7905 Labmate Pro Starter Kit with LMP10, LMP20, LMP200 & LMP1000 Pipettors and Accessories
| Mfr: 7905| Item: 2455221
Designed to make clinically demanding applications feel physically effortless
£755.02GBP / Each
£906.02GBP (incl VAT)
£755.02GBP / Each
£906.02GBP (incl VAT)
Product Description
Includes
Pipettor models 24551-95, -96, -98, -99, Plexi 4-position stand 96 tips 10 μl, 96 tips 200 μl, 96 tips 1000 μl, calibration tool x 4, and pipette hanger x 4.
Key Features
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • 3 year warranty
More About this Item
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed, Yellow, and Blue
  • DescriptionLabmate Pro Starter Kit with LMP10, LMP20, LMP200 & LMP1000 Pipettors and Accessories