HTL 4053 Discovery Comfort D20 Single Channel Pipettor with Colored Shaft, Yellow; 2 to 20 µL
Representative image only.
HTL 4053 Discovery Comfort D20 Single Channel Pipettor with Colored Shaft, Yellow; 2 to 20 µL
| Mfr: 4053| Item: 2455147
Comfort, safety, precision and value make this variable-volume pipette series quite a discovery
£209.61GBP / Each
£251.53GBP (incl VAT)
£209.61GBP / Each
£251.53GBP (incl VAT)
Product Description
Key Features
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision. Multichannel models feature a revolutionary suspension system which allows the shafts to retract slightly when they are pressed against a row of pipette tips.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)20
  • Min Volume (µL)2
  • Accuracy (%)0.8
  • Precision (%)0.3
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionDiscovery Comfort D20 Single Channel Pipettor with Colored Shaft, Yellow; 2 to 20 µL