HTL 4051 Discovery Comfort D2 Single Channel Pipettor with Colored Shaft, Red; 0.2 to 2 µL
Representative image only.
HTL 4051 Discovery Comfort D2 Single Channel Pipettor with Colored Shaft, Red; 0.2 to 2 µL
| Mfr: 4051| Item: 2455145
Comfort, safety, precision and value make this variable-volume pipette series quite a discovery
£230.83GBP / Each
£277.00GBP (incl VAT)
£230.83GBP / Each
£277.00GBP (incl VAT)
Product Description
Key Features
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision. Multichannel models feature a revolutionary suspension system which allows the shafts to retract slightly when they are pressed against a row of pipette tips.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)2
  • Min Volume (µL)0.2
  • Accuracy (%)1.5
  • Precision (%)0.7
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed
  • DescriptionDiscovery Comfort D2 Single Channel Pipettor with Colored Shaft, Red; 0.2 to 2 µL