EPC3155 Cubitainer, LDPE, 1 Liter; 128/Cs
Representative image only.
EPC3155 Cubitainer, LDPE, 1 Liter; 128/Cs
| Mfr: EPC3155| Item: 3520490
Collapsible containers nest for efficient storage
£513.70GBP / Case of 128
£616.44GBP (incl VAT)
£513.70GBP / Case of 128
£616.44GBP (incl VAT)
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
3 Month Warranty
Specifications
  • Capacity (L)1
  • MaterialLDPE
  • DescriptionCubitainer, LDPE, 1 Liter; 128/Cs