EPC3030 Cubitainer, LDPE, 1 Quart; 12/Cs
Representative image only.
EPC3030 Cubitainer, LDPE, 1 Quart; 12/Cs
| Mfr: EPC3030| Item: 3520479
Collapsible containers nest for efficient storage
£50.16GBP / Case of 12
£60.19GBP (incl VAT)
£50.16GBP / Case of 12
£60.19GBP (incl VAT)
Buy It Together
Current Item
EPC3030 Cubitainer, LDPE, 1 Quart; 12/Cs
EPC3030 Cubitainer, LDPE, 1 Quart; 12/Cs
3520479
£50.16GBP / Case of 12
£60.19GBP (incl VAT)
+
LDPE Coupling Cap, 38 mm for 06061-12 and Collapsible LDPE Containers
0610080
£14.36GBP / Pkg of 6
£17.23GBP (incl VAT)
=
Total Price 64.52 ADD 2 ITEMS TO CART
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Natural LDPE containers come assembled with LDPE-lined size 38 caps.
3 Month Warranty
Specifications
  • Capacity (mL)946.353
  • Capacity (L)0.946
  • Capacity (oz)32
  • Capacity (Gal)0.25
  • MaterialLow Density Polyethyene (LDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, LDPE, 1 Quart; 12/Cs