EPC3000 Cubitainer, LDPE, 20 Liter; 36/Cs
Representative image only.
EPC3000 Cubitainer, LDPE, 20 Liter; 36/Cs
| Mfr: EPC3000| Item: 3520488
Collapsible containers nest for efficient storage
£273.69GBP / Case of 36
£328.43GBP (incl VAT)
£273.69GBP / Case of 36
£328.43GBP (incl VAT)
Buy It Together
Current Item
EPC3000 Cubitainer, LDPE, 20 Liter; 36/Cs
EPC3000 Cubitainer, LDPE, 20 Liter; 36/Cs
3520488
£273.69GBP / Case of 36
£328.43GBP (incl VAT)
+
LDPE Coupling Cap, 38 mm for 06061-12 and Collapsible LDPE Containers
0610080
£14.36GBP / Pkg of 6
£17.23GBP (incl VAT)
=
Total Price 288.05 ADD 2 ITEMS TO CART
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Natural LDPE containers come assembled with LDPE-lined size 38 caps.
3 Month Warranty
Specifications
  • Capacity (mL)20000
  • Capacity (L)20
  • Capacity (oz)676
  • Capacity (Gal)5
  • MaterialLow Density Polyethyene (LDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, LDPE, 20 Liter; 36/Cs