Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 4 mL Nitric Acid Preserved, 500 mL; 24/CS
Representative image only.
Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 4 mL Nitric Acid Preserved, 500 mL; 24/CS
Environmental Express | Item: 3521044
Level 1 bottles are certified and come with a certificate of analysis
£96.02GBP / Case of 24
£115.22GBP (incl VAT)
£96.02GBP / Case of 24
£115.22GBP (incl VAT)
Product Description
Key Features
  • Preserved and unpreserved bottles available
More About this Item
These HDPE bottles are Level 1 certified to meet the EPA specifications and guidance for contaminant free sample collection containers.
Specifications
  • Capacity (mL)500
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Level Number1
  • Special Features4 mL nitric acid preserved
  • Neck Inside Diameter (ID) (mm)53
  • DescriptionLevel 1 Certified Natural Color Bottle, Round, Wide Mouth, 4 mL Nitric Acid Preserved, 500 mL, 24/Case