Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 2 mL Nitric Acid Preserved, 250 mL; 24/CS
Representative image only.
Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 2 mL Nitric Acid Preserved, 250 mL; 24/CS
Environmental Express | Item: 3520956
Level 1 bottles are certified and come with a certificate of analysis
£52.28GBP / Case of 24
£62.74GBP (incl VAT)
£52.28GBP / Case of 24
£62.74GBP (incl VAT)
Product Description
Key Features
  • Preserved and unpreserved bottles available
More About this Item
These HDPE bottles are Level 1 certified to meet the EPA specifications and guidance for contaminant free sample collection containers.
Specifications
  • Capacity (mL)250
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Level Number1
  • Special Features2 mL nitric acid preserved
  • Neck Inside Diameter (ID) (mm)43
  • DescriptionLevel 1 Certified Natural Color Bottle, Round, 2 mL Nitric Acid Preserved, 250 mL, 24/Case