Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 0.6 mL Sulfuric Acid Preserved 125 mL; 48/CS
Representative image only.
Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 0.6 mL Sulfuric Acid Preserved 125 mL; 48/CS
Environmental Express | Item: 3520923
Level 1 bottles are certified and come with a certificate of analysis
£71.11GBP / Case of 48
£85.33GBP (incl VAT)
£71.11GBP / Case of 48
£85.33GBP (incl VAT)
Product Description
Key Features
  • Preserved and unpreserved bottles available
More About this Item
These HDPE bottles are Level 1 certified to meet the EPA specifications and guidance for contaminant free sample collection containers.
Specifications
  • Capacity (mL)125
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Special Features0.6 mL sulfuric acid preserved
  • Level Number1
  • Neck Inside Diameter (ID) (mm)38
  • DescriptionLevel 1 Certified Natural Color Bottle, Round, 0.6 mL Sulfuric Acid Preserved, 125 mL, 48/Case