Environmental Express Wide-Mouth Amber Glass Bottle, Level 1, 60 mL, PC Cap; 12/CS
Representative image only.
Environmental Express Wide-Mouth Amber Glass Bottle, Level 1, 60 mL, PC Cap; 12/CS
Environmental Express | Item: 3520402
Larger cap for easier sampling
£26.77GBP / Case of 12
£32.12GBP (incl VAT)
£26.77GBP / Case of 12
£32.12GBP (incl VAT)
Buy It Together
Current Item
Environmental Express Wide-Mouth Amber Glass Bottle, Level 1, 60 mL, PC Cap; 12/CS
Environmental Express Wide-Mouth Amber Glass Bottle, Level 1, 60 mL, PC Cap; 12/CS
3520402
£26.77GBP / Case of 12
£32.12GBP (incl VAT)
+
Cole-Parmer Pre-Cleaned EPA Narrow-Mouth Bottle, Amber Glass, 125mL (4 oz); 24/CS
9954030
£63.64GBP / Case of 24
£76.37GBP (incl VAT)
+
Cole-Parmer Pre-Cleaned EPA Wide-Mouth Bottle, Amber Glass, 500mL (17 oz); 12/CS
9954055
£54.26GBP / Case of 12
£65.11GBP (incl VAT)
=
Total Price 144.67 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from two cleanliness levels—Level 1 include a Certificate of Analysis where Level 3 are perfect for general use
  • F217-lined caps provide a tight seal with some chemical resistance
  • Select amber glass bottles for light-sensitive samples
  • 3 month warranty
  • Model number APC1252
More About this Item
Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory. Included caps are Polycone and lined with PTFE.
3 Month Warranty
Specifications
  • Capacity (mL)60
  • Capacity (L)0.06
  • Capacity (oz)2
  • MaterialGlass
  • Cap MaterialPTFE-lined Polycone
  • Cap / Stopper Size33 mm
  • Bottle ColorAmber
  • Level Number1
  • Special FeaturesPC cap
  • AutoclavableNo
  • DescriptionWide-Mouth Amber Glass Bottle, Level 1, 60 mL, PC Cap; 12/Cs