Environmental Express R2200 PVC "Sticky" Planchette; 100/Pk
Representative image only.
Environmental Express R2200 PVC "Sticky" Planchette; 100/Pk
Environmental Express | Item: 3520007
Useful planchettes make spectroscopy easy
£84.62GBP / Pkg of 100
£101.54GBP (incl VAT)
£84.62GBP / Pkg of 100
£101.54GBP (incl VAT)
Product Description
Key Features
  • For alpha particle spectroscopy
  • Incorporates contact adhesive
  • Model number R2200
More About this Item
Place collection filter on the adhesive to keep the filter flat for optimized analysis and repeatability.
Specifications
  • MaterialPVC
  • Diameter (mm)28.6
  • Diameter (cm)2.86
  • DescriptionPVC "Sticky" Planchette, 100/Pk