Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Nitrate
Representative image only.
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Nitrate
Environmental Express | Item: 3581230
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
£377.75GBP / Each
£453.30GBP (incl VAT)
£377.75GBP / Each
£453.30GBP (incl VAT)
Buy It Together
Current Item
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Nitrate
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Nitrate
3581230
£377.75GBP / Each
£453.30GBP (incl VAT)
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
£128.06GBP / Each
£153.67GBP (incl VAT)
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
£407.27GBP / Each
£488.72GBP (incl VAT)
+
Environmental Express Oakton pH 700 Benchtop Meter with Probe Stand
3541912
£879.40GBP / Each
£1,055.28GBP (incl VAT)
+
Environmental Express Oakton pH/Ion 700 Ion 700 Benchtop Meter with electrode stand
3541922
£853.85GBP / Each
£1,024.62GBP (incl VAT)
+
Environmental Express EC 2000 Benchtop Meter Kit
3541960
£1,108.39GBP / Each
£1,330.07GBP (incl VAT)
+ + =
Total Price 4539.11 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction, membrane
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeNitrate
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Double-Juntion, Epoxy; Nitrate