Environmental Express HDPE Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 8 mL Nitric Acid Preserved, 1000 mL; 12/CS
Representative image only.
Environmental Express HDPE Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 8 mL Nitric Acid Preserved, 1000 mL; 12/CS
Environmental Express | Item: 3521048
Level 1 bottles are certified and come with a certificate of analysis
£51.32GBP / Case of 12
£61.58GBP (incl VAT)
£51.32GBP / Case of 12
£61.58GBP (incl VAT)
Product Description
Key Features
  • Preserved and unpreserved bottles available
More About this Item
These HDPE bottles are Level 1 certified to meet the EPA specifications and guidance for contaminant free sample collection containers.
Specifications
  • Capacity (mL)1000
  • Capacity (L)1
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Level Number1
  • Special Features8 mL nitric acid preserved
  • Neck Inside Diameter (ID) (mm)63
  • DescriptionLevel 1 Certified Natural Color Bottle, Round, Wide Mouth, 8 mL Nitric Acid Preserved, 1 L, 12/Case