Environmental Express Distillation Head, for Cyanide Glassware Set
Representative image only.
Environmental Express Distillation Head, for Cyanide Glassware Set
Environmental Express | Item: 3520964
Replacement components for the MicroBlock systems
£181.70GBP / Each
£218.04GBP (incl VAT)
£181.70GBP / Each
£218.04GBP (incl VAT)
Product Description
Key Features
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 3 month warranty
More About this Item
Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.
3 Month Warranty
Specifications
  • DescriptionDistillation Head, for Cyanide Glassware Set