Environmental Express Dairy-Style Jug, HDPE, Level 3, 1 gal; 20/Cs
Representative image only.
Environmental Express Dairy-Style Jug, HDPE, Level 3, 1 gal; 20/Cs
Environmental Express | Item: 3520679
Grab and pour with easy-grip handle
£29.22GBP / Case of 20
£35.06GBP (incl VAT)
£29.22GBP / Case of 20
£35.06GBP (incl VAT)
Product Description
Key Features
  • Leakproof design keeps contents safe inside
  • Familiar and easy pouring with the milk-type jug design
  • Pre-cleaned for immediate use
  • 3 month warranty
  • Model number BPC3042
More About this Item
Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE. Level 3 pre-cleaned containers meet EPA guidelines, but do not ship with a Certificate of Analysis.
3 Month Warranty
Specifications
  • Capacity (mL)3845
  • Capacity (L)3.845
  • Capacity (Gal)1
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialLDPE-lined Polypropylene
  • Cap / Stopper Size38 mm
  • Cap ColorWhite
  • Bottle ColorNatural
  • Level Number3
  • AutoclavableNo
  • DescriptionDairy-Style Jug, HDPE, 1 gal; 20/Cs