Environmental Express BOD: SimpleWater (Nutrient Buffer Solution), 300mL, 36/Box

Representative image only.

Environmental Express BOD: SimpleWater (Nutrient Buffer Solution), 300mL, 36/Box

Environmental Express | Item: 5320001

No mess--simply pour contents into your container and add water

£24.36GBP / Pkg of 36
£29.23GBP (incl VAT)

£24.36GBP / Pkg of 36
£29.23GBP (incl VAT)
Product Description

Key Features

  • Range of five standard concentrations to make a volume of 300 mL to 19 L
  • Borosilicate glass vials with leakproof screw caps
  • 3 month warranty

More About this Item

BOD dilution water is composed of a variety of nutrients that provide optimal growth media for bacteria in BOD tests. SimpleWater nutrient buffer solutions are concentrates of the four nutrient solutions conveniently combined into one vial. Simply dilute the contents of vial to the specified volume, and you've got your BOD dilution water!

Available in five standard concentrations, borosilicate glass vials with leakproof screwcaps. Vials in pack of 36.

3 Month Warranty
Specifications
  • ClassBOD Dilution Water Concentrate
  • SynonymsBOD dilution water
  • FormLiquid
  • Volume (mL)300
  • Weight (mL)300
  • Packaging300 mL For 300 mL dilution
  • DOT InformationNot regulated by DOT
  • CAS Number12125-02-9, 10035-04-8, 10025-77-1, 10034-99-8, 7758-11-4, 7778-77-0, and 7558-79-4
  • DescriptionBOD: SimpleWater (Nutrient Buffer Solution), 300mL, 36/Box
  • Warranty3 month
Reviews
Q&A