Environmental Express BOD Bottle Storage Rack, Epoxy-Coated Steel, 12-place
Representative image only.
Environmental Express BOD Bottle Storage Rack, Epoxy-Coated Steel, 12-place
Environmental Express | Item: 5320022
Safe and convenient organization of BOD bottles
£89.38GBP / Each
£107.26GBP (incl VAT)
£89.38GBP / Each
£107.26GBP (incl VAT)
Product Description
Includes
Racks do not include BOD bottles.
Key Features
  • Stackable
  • Choice of polypropylene or steel construction
  • 3 month warranty
  • Model number D1052
More About this Item
Polypropylene racks can be autoclaved up to 121°C. Steel racks are more durable but cannot be autoclaved.
These stackable rugged wire racks are plastic-coated to avoid corrosion.
3 Month Warranty
Specifications
  • MaterialEpoxy-coated steel
  • Compatible With300 mL BOD bottles
  • Length (InchShort)13
  • Height (in)8
  • Width (InchShort)10
  • DescriptionBOD Bottle Storage Rack, Epoxy-coated Steel, 300 mL, 12-place; Each