Cole-Parmer VapLock™ Solvent Waste Manifold, PP, DIN 50, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Representative image only.
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, DIN 50, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Cole-Parmer | Item: 1201889
Connect solvent waste manifold caps to waste containers for a secure closed system
£146.06GBP / Each
£175.27GBP (incl VAT)
£146.06GBP / Each
£175.27GBP (incl VAT)
Buy It Together
Current Item
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, DIN 50, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, DIN 50, 1x1/2" NPT, 3x1/4" NPT; 1/EA
1201889
£146.06GBP / Each
£175.27GBP (incl VAT)
+ + +
Cole-Parmer VapLock™ Exhaust Filter, 75 g activated carbon, 1/4" NPT(F); 1/ea
1201979
£60.86GBP / Each
£73.03GBP (incl VAT)
+
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 2 Luer Ports without Valve, Blue
2194311
£27.71GBP / Each
£33.25GBP (incl VAT)
=
Total Price 656.57 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from polypropylene or PTFE construction
  • DIN 50 connection
  • Polypropylene construction
  • 4 ports
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Bottle and carboy caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
Specifications
  • MaterialPolypropylene(PP)
  • Cap / Stopper SizeDIN 50
  • Cap ColorWhite
  • Special Features1 x 1/2" NPT(F), 3 x 1/4" NPT(F)
  • DescriptionSolvent Waste Manifold, PP, DIN 50, 1x1/2" NPT, 3x1/4" NPT; 1/EA