Cole-Parmer Testing Sieve, SS Frame/SS Wire, 12" OD, Half Height, No. 400
Representative image only.
Cole-Parmer Testing Sieve, SS Frame/SS Wire, 12" OD, Half Height, No. 400
Cole-Parmer | Item: 5999524
Get more accurate results
£408.60GBP / Each
£490.32GBP (incl VAT)
for an additional
£151.83GBP / Each
£182.20GBP (incl VAT)
for an additional
£201.86GBP / Each
£242.23GBP (incl VAT)
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
£408.60GBP / Each
£490.32GBP (incl VAT)
Buy It Together
Current Item
Cole-Parmer Testing Sieve, SS Frame/SS Wire, 12" OD, Half Height, No. 400
Cole-Parmer Testing Sieve, SS Frame/SS Wire, 12" OD, Half Height, No. 400
5999524
£408.60GBP / Each
£490.32GBP (incl VAT)
+
InnoCal NIST-Traceable Calibration; Sieve
1705050
£151.83GBP / Each
£182.20GBP (incl VAT)
+
W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
5998696
£28.71GBP / Each
£34.45GBP (incl VAT)
+
W.S. Tyler 8532 Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
5999690
£146.23GBP / Each
£175.48GBP (incl VAT)
=
Total Price 735.37 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
  • 1 year warranty
More About this Item
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)400
  • Tyler Standard Screen Size (mesh)400
  • Sieve Opening (mm)0.037
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionTesting Sieve, SS Frame/SS Wire, 12" OD, Half height, No. 400