Cole-Parmer SamplePrep 3650 Vertrel™ XF Liquid Fluorocarbon Grinding Aid, 1 Quart
Representative image only.
Cole-Parmer SamplePrep 3650 Vertrel™ XF Liquid Fluorocarbon Grinding Aid, 1 Quart
Cole-Parmer SamplePrep | Mfr: 3650| Item: 0457623
Prepare samples for reproducible and contamination-free results
£302.00GBP / Each
£362.40GBP (incl VAT)
£302.00GBP / Each
£362.40GBP (incl VAT)
Product Description
Key Features
  • Select binders from powder, tablet, or liquid form depending on your application
  • Ideal for grinding samples such as cement, rock, and clinker
  • 1 quart bottle
More About this Item
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
Liquid fluorocarbon grinding aid coats the particles then evaporates after grinding without leaving any residue.
Specifications
  • MaterialLiquid
  • DescriptionVertrel™ XF Liquid Fluorocarbon Grinding Aid, 1 Quart