Cole-Parmer SamplePrep 3644T-1500 UltraBind® Binder Tablets, 0.5 g Each, 1500 Tablets
Representative image only.
Cole-Parmer SamplePrep 3644T-1500 UltraBind® Binder Tablets, 0.5 g Each, 1500 Tablets
Cole-Parmer SamplePrep | Mfr: 3644T-1500| Item: 0457752
Prepare samples for reproducible and contamination-free results
£471.00GBP / Each
£565.20GBP (incl VAT)
£471.00GBP / Each
£565.20GBP (incl VAT)
Product Description
Key Features
  • Select binders from powder, tablet, or liquid form depending on your application
  • 1500 tablets
More About this Item
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
To use, add tablets to sample (~10% by weight) and blend. It can also be used as a sample disk matrix with our sleeve-and-plunger set 04577-38 or 04577-45. Does not cake in the bottle during storage.
Specifications
  • MaterialTablet
  • DescriptionUltraBind® Binder Tablets, 0.5 g Each, 1500 Tablets