Cole-Parmer Nipple Fitting, Straight Reducer, Nylon, 1/2" x 1/8"; 10/PK
Representative image only.
Cole-Parmer Nipple Fitting, Straight Reducer, Nylon, 1/2" x 1/8"; 10/PK
Cole-Parmer | Item: 5014393
High-purity fittings deliver exceptional chemical resistance, durability, and compliance for critical fluid handling in food, bioprocess, and bioresearch applications
Call For Price
Call For Price
Product Description
Key Features
  • Injection molded from high-purity, semi-opaque fluoropolymer
  • Provides excellent chemical and corrosion resistance across harsh environments
  • Free from animal-derived materials, plasticizers, phthalates, and latex
  • Stable across a wide temperature range: -35 to 135°C (-31 to 275°F)
  • Supports ultrasonic welding for downstream processing flexibility
  • Sterilize by autoclave, gamma irradiation, or EtO
More About this Item
Engineered from a high-purity thermoplastic fluoropolymer, these PVDF fittings meet the rigorous demands of food, beverage, bioprocess, and bioresearch applications. This advanced material combines outstanding chemical resistance and corrosion protection with exceptional durability, maintaining performance across a wide temperature range from -35 to 135°C (-31 to 275°F).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Chemical ResistanceResistant to fuels, lubricants and many chemicals; vulnerable to phenols, strong acids, and oxidizing agents.
  • Connection TypeNPT(M) x NPT(M)
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/2
  • Thread Size NPT(M) (Port 2) (InchShort)1/8
  • Flow PatternStraight
  • ColorNatural
  • Regulatory ApprovalsRoHS
  • Quantity10
  • DescriptionCole-Parmer Reduce Nipple Fittings, 1/2" x 1/8" Nylon, 10PK