Cole-Parmer Hepatitis Delta Virus, With Mastermix
Representative image only.
Cole-Parmer Hepatitis Delta Virus, With Mastermix
Cole-Parmer | Item: 9394243
Quickly and accurately detect the presence of human hepatitis infection
Call For Price
Call For Price
Product Description
Includes
Onestep mastermix (RNA transcriptase), lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control RNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
Specifications
  • Test TypeRNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • Binomial NameHepatitis Delta Virus
  • DescriptionHepatitis Delta Virus, With Mastermix