Cole-Parmer DNA Genesig NOS terminator in GM crops, with mastermix
Representative image only.
Cole-Parmer DNA Genesig NOS terminator in GM crops, with mastermix
Cole-Parmer | Item: 9394026
Screen for genetically modified organisms (GMOs) in food, feed, and seeds
Call For Price
Call For Price
Product Description
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Kits with mastermix contain all the necessary components for fast and accurate results. To use your own mastermix, order kits without mastermix.
Specifications
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • DescriptionGenesig NOS terminator in GM crops, with mastermix