Cole-Parmer DNA Genesig CaMV 35S promoter in GM crops, without mastermix
Representative image only.
Cole-Parmer DNA Genesig CaMV 35S promoter in GM crops, without mastermix
Cole-Parmer | Item: 9394008
Screen for genetically modified organisms (GMOs) in food, feed, and seeds
Call For Price
Call For Price
Product Description
Includes
Lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Kits with mastermix contain all the necessary components for fast and accurate results. To use your own mastermix, order kits without mastermix.
Specifications
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedNo
  • DescriptionGenesig CaMV 35S promoter in GM crops, without mastermix