Cole-Parmer Coplin Slide Staining Jar with Domed Cap, Polypropylene, Holds 10 Slides; 12/Pk
Representative image only.
Cole-Parmer Coplin Slide Staining Jar with Domed Cap, Polypropylene, Holds 10 Slides; 12/Pk
Cole-Parmer | Item: 7890744
Ideal for staining slides
£29.04GBP / Pkg of 12
£34.85GBP (incl VAT)
£29.04GBP / Pkg of 12
£34.85GBP (incl VAT)
Product Description
Key Features
  • Constructed of durable polypropylene
  • Jar is designed to hold up to 10 standard 25 x 75mm (1 x 3”) slides back to back
  • Autoclavable
More About this Item
Cole-Parmer® Essentials Coplin staining jar features a domed, shallow thread screw cap to protect slides from outside elements and reduce reagent evaporation. Jar interior is grooved to hold ten 3” x 1” slides back-to-back in an upright position.
Specifications
  • Capacity (Number of slides)10
  • ColorWhite
  • DescriptionCoplin Slide Staining Jar with Domed Cap, Polypropylene, Holds 10 Slides; 12/Pk