Cole-Parmer Chlamydiaceae (All Species), Without Mastermix
Representative image only.
Cole-Parmer Chlamydiaceae (All Species), Without Mastermix
Cole-Parmer | Item: 9394655
Screen for systemic microbial pathogens quickly to prevent economic loss
Call For Price
Call For Price
Product Description
Includes
Lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Prevent economic loss, especially in the dairy and agricultural industry, using these microbial detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
Specifications
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedNo
  • Binomial NameChlamydiaceae (All Species)
  • DescriptionChlamydiaceae (All Species), Without Mastermix